Assay Details
| Gene Symbol: Dlg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109752.1 | NP_001103222.1 | 12 - 13 | 1785 | 77 | |
| NM_007864.3 | NP_031890.1 | 12 - 13 | 1794 | 77 | ||
| GenBank mRNA | BC014807.1 | - | 12 - 13 | 1530 | 4501403 | 77 |
| D50621.1 | - | 12 - 13 | 1536 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dlg4
Genomic Map