Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100008132.mm.1 |
NM_001109752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008132.mm.1 |
NM_001109752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008132.mm.1 |
NM_007864.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008132.mm.1 |
NM_007864.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008132.mm.1 |
TC1100000937.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008129.mm.1 |
NM_001109752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008129.mm.1 |
NM_001109752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008129.mm.1 |
NM_007864.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008129.mm.1 |
NM_007864.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008129.mm.1 |
TC1100000937.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008130.mm.1 |
NM_001109752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008130.mm.1 |
NM_001109752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008130.mm.1 |
NM_007864.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008130.mm.1 |
NM_007864.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008130.mm.1 |
TC1100000937.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008135.mm.1 |
NM_001109752.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008135.mm.1 |
NM_001109752.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008135.mm.1 |
NM_007864.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008135.mm.1 |
NM_007864.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008135.mm.1 |
TC1100000937.mm.1 |
MTA TC |