Assay Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153788.3 | NP_722483.2 | 6 - 7 | 730 | 83 | |
| GenBank mRNA | AK133473.1 | - | 4 - 5 | 1205 | 83 | |
| AK138736.1 | - | 6 - 7 | 722 | 83 | ||
| AK154210.1 | - | 6 - 7 | 695 | 83 | ||
| AK154449.1 | - | 6 - 7 | 733 | 83 | ||
| AK155001.1 | - | 6 - 7 | 751 | 83 | ||
| BC037481.1 | - | 1 - 2 | 357 | 4460639 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037481.1 transcripts.
Set Membership: |
|

Acap1
Genomic Map