Assay Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153788.3 | NP_722483.2 | 7 - 8 | 774 | 80 | |
| GenBank mRNA | AK133473.1 | - | 5 - 6 | 1249 | 80 | |
| AK138736.1 | - | 7 - 8 | 766 | 80 | ||
| AK154210.1 | - | 7 - 8 | 739 | 80 | ||
| AK154449.1 | - | 7 - 8 | 777 | 80 | ||
| AK155001.1 | - | 7 - 8 | 795 | 80 | ||
| BC037481.1 | - | 2 - 3 | 401 | 4460639 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC037481.1 transcripts.
Set Membership: |
|

Acap1
Genomic Map