Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 5 - 6 | 420 | 85 | |
| GenBank mRNA | AF105217.1 | - | 5 - 6 | 395 | 85 | |
| AJ249341.2 | - | 5 - 6 | 398 | 85 | ||
| AK087557.1 | - | 5 - 6 | 421 | 85 | ||
| AK154160.1 | - | 5 - 6 | 414 | 85 | ||
| AK187899.1 | - | 1 - 2 | 49 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map