Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 7 - 8 | 572 | 73 | |
| NR_045642.1 | 6 - 7 | 498 | 73 | |||
| GenBank mRNA | AF105217.1 | - | 7 - 8 | 547 | 73 | |
| AJ249341.2 | - | 7 - 8 | 550 | 73 | ||
| AK041651.1 | - | 7 - 8 | 563 | 73 | ||
| AK051286.1 | - | 6 - 7 | 492 | 73 | ||
| AK087557.1 | - | 7 - 8 | 573 | 73 | ||
| AK154160.1 | - | 7 - 8 | 566 | 73 | ||
| AK178795.1 | - | 3 - 4 | 197 | 73 | ||
| AK180350.1 | - | 2 - 3 | 124 | 73 | ||
| AK187899.1 | - | 3 - 4 | 201 | 73 | ||
| AK193655.1 | - | 3 - 4 | 197 | 73 | ||
| AK196494.1 | - | 3 - 4 | 197 | 73 | ||
| AK216095.1 | - | 3 - 4 | 197 | 73 | ||
| BC019982.1 | - | 7 - 8 | 564 | 4159742 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map