Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 8 - 9 | 657 | 60 | |
| NR_045642.1 | 7 - 8 | 583 | 60 | |||
| GenBank mRNA | AF105217.1 | - | 8 - 9 | 632 | 60 | |
| AJ249341.2 | - | 8 - 9 | 635 | 60 | ||
| AK041651.1 | - | 8 - 9 | 648 | 60 | ||
| AK051286.1 | - | 7 - 8 | 577 | 60 | ||
| AK087557.1 | - | 8 - 9 | 658 | 60 | ||
| AK154160.1 | - | 8 - 9 | 651 | 60 | ||
| BC019982.1 | - | 8 - 9 | 649 | 4159742 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map