Assay Details
| Gene Symbol: Aldh1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153543.2 | NP_705771.2 | 6 - 7 | 915 | 70 | |
| GenBank mRNA | AK052361.1 | - | 6 - 7 | 806 | 70 | |
| AK086413.1 | - | 6 - 7 | 70 | |||
| AK141954.1 | - | 6 - 7 | 877 | 70 | ||
| AK215056.1 | - | 3 - 4 | 228 | 70 | ||
| BC034531.1 | - | 6 - 7 | 837 | 4188055 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1l2
Genomic Map