Assay Details
| Gene Symbol: Dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013380.2 | NP_001013398.2 | 3 - 4 | 546 | 137 | |
| GenBank mRNA | AK050575.1 | - | 3 - 4 | 515 | 137 | |
| AK140743.1 | - | 3 - 4 | 314 | 137 | ||
| AK147899.1 | - | 3 - 4 | 313 | 137 | ||
| AK152157.1 | - | 3 - 4 | 311 | 137 | ||
| AK161434.1 | - | 3 - 4 | 408 | 137 | ||
| AK161494.1 | - | 3 - 4 | 408 | 137 | ||
| BC058645.1 | - | 3 - 4 | 504 | 6405596 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dync1li2
Genomic Map