Assay Details
| Gene Symbol: Dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013380.2 | NP_001013398.2 | 9 - 10 | 1345 | 123 | |
| GenBank mRNA | AK147899.1 | - | 9 - 10 | 1112 | 123 | |
| AK152157.1 | - | 9 - 10 | 1110 | 123 | ||
| AK161434.1 | - | 9 - 10 | 1207 | 123 | ||
| AK161494.1 | - | 9 - 10 | 1207 | 123 | ||
| AK207934.1 | - | 1 - 2 | 33 | 123 | ||
| BC058645.1 | - | 9 - 10 | 1303 | 6405596 | 123 | |
More Information
Set Membership: |
|

Dync1li2
Genomic Map