Assay Details
| Gene Symbol: Tradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033161.2 | NP_001028333.1 | 1 - 2 | 34 | 66 | |
| GenBank mRNA | AK156540.1 | - | 1 - 2 | 34 | 66 | |
| BC132607.1 | - | 1 - 2 | 30 | 40130884 | 66 | |
| BC138642.1 | - | 1 - 2 | 30 | 8861662 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156540.1, BC132607.1, BC138642.1, NM_001033161.2 transcripts.
Set Membership: |
|

Tradd
Genomic Map