Assay Details
| Gene Symbol: Tradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033161.2 | NP_001028333.1 | 3 - 4 | 467 | 75 | |
| GenBank mRNA | AK018592.1 | - | 3 - 4 | 481 | 75 | |
| AK156540.1 | - | 3 - 4 | 467 | 75 | ||
| BC132607.1 | - | 3 - 4 | 463 | 40130884 | 75 | |
| BC138642.1 | - | 3 - 4 | 463 | 8861662 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein heterooligomerization
positive regulation of hair follicle development
extrinsic apoptotic signaling pathway
signal transducer activity
tumor necrosis factor receptor binding
protein binding
kinase binding
protein complex binding
identical protein binding
binding, bridging
death domain binding
apoptotic process
signal transduction
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein heterooligomerization
positive regulation of hair follicle development
extrinsic apoptotic signaling pathway
signal transducer activity
tumor necrosis factor receptor binding
protein binding
kinase binding
protein complex binding
identical protein binding
binding, bridging
death domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132555 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763696 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143953 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135933 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109194 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132579 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06626016 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109194 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135933 | - | - |
| Whole Mouse Genome G4122A | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288505 | BC028542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288505 | AK018592.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109194 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135933 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143953 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06846131 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06846120 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132555 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07231039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581183 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439910_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429117_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452622_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581181 | - | - |
| Mouse Genome 430 2.0 Array | 1452622_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1439910_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452622_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1439910_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429117_at | - | - |
| Mouse Genome 430 2.0 Array | 1429117_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71609_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71609_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021588.mm.1 | NM_001033161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021588.mm.1 | NM_001033161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021588.mm.1 | TC0800002812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021584.mm.1 | NM_001033161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021584.mm.1 | NM_001033161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021584.mm.1 | TC0800002812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021589.mm.1 | NM_001033161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021589.mm.1 | NM_001033161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021589.mm.1 | TC0800002812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021585.mm.1 | NM_001033161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021585.mm.1 | NM_001033161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021585.mm.1 | TC0800002812.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021586.mm.1 | NM_001033161.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021586.mm.1 | NM_001033161.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021586.mm.1 | TC0800002812.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1452622_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452622_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429117_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429117_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439910_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439910_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 395830 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2505100 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2505100 | - | - |

Tradd
Genomic Map