Assay Details
| Gene Symbol: 4931428F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166394.1 | NP_001159866.1 | 1 - 2 | 121 | 63 | |
| GenBank mRNA | AK016481.1 | - | 1 - 2 | 456 | 63 | |
| BC059045.1 | - | 1 - 2 | 56 | 6816353 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016481.1, BC059045.1, NM_001166394.1 transcripts.
Set Membership: |
|

4931428F04Rik
Genomic Map