Assay Details
| Gene Symbol: Tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026481.3 | NP_080757.1 | 2 - 3 | 340 | 79 | |
| GenBank mRNA | AK009771.1 | - | 2 - 3 | 304 | 79 | |
| AK012437.1 | - | 2 - 3 | 308 | 79 | ||
| AK155018.1 | - | 2 - 3 | 311 | 79 | ||
| AK160507.1 | - | 2 - 3 | 311 | 79 | ||
| BC010788.1 | - | 2 - 3 | 297 | 4216285 | 79 | |
| BC116622.1 | - | 1 - 2 | 132 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tppp3
Genomic Map