Assay Details
| Gene Symbol: Tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026481.3 | NP_080757.1 | 3 - 4 | 492 | 65 | |
| GenBank mRNA | AK009771.1 | - | 3 - 4 | 456 | 65 | |
| AK012437.1 | - | 3 - 4 | 460 | 65 | ||
| AK155018.1 | - | 3 - 4 | 463 | 65 | ||
| AK160507.1 | - | 3 - 4 | 463 | 65 | ||
| BC010788.1 | - | 3 - 4 | 449 | 4216285 | 65 | |
| BC116622.1 | - | 2 - 3 | 284 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133998 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 693899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461665 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141557 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134870 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142020 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26134002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133980 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142020 | - | - |
| Whole Mouse Genome G4122A | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK155018.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK160507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK009771.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AK012437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246924 | AW494372.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126913 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120704 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134870 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141557 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06842490 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421420 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07421396 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133999 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26134000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26134002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26134008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26134006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07151607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07421402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26134002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07421420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07421398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06842490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26134004 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581268 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416713_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581266 | - | - |
| Mouse Genome 430 2.0 Array | 1416713_at | - | - |
| Mouse Genome 430 2.0 Array | 1416713_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67971_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67971_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021713.mm.1 | NM_026481.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021713.mm.1 | NM_026481.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021713.mm.1 | TC0800002822.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021711.mm.1 | NM_026481.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021711.mm.1 | NM_026481.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021711.mm.1 | TC0800002822.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416713_PM_at | - | - |
| Mouse Expression Set 430 | 1416713_at | - | - |
| Murine Genome U74v2 Set | 95108_PM_at | - | - |
| Murine Genome U74v2 Set | 95108_at | - | - |
| Murine Genome U74v2 Set | 171613_at | - | - |
| Murine Genome U74v2 Set | 171613_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416713_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416713_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416713_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416713_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2470148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2470148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229878 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229878 | - | - |
| mouse-6 v1.1 (Illumina) | scl34402.5.1_27-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34402.5.1_27-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2655929 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2655929 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34402.5.1_27-S | - | - |

Tppp3
Genomic Map