Assay Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081333.1 | NP_001074802.1 | 10 - 11 | 1488 | 83 | |
| GenBank mRNA | AK131125.1 | - | 8 - 9 | 1766 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhg4
Genomic Map