Assay Details
| Gene Symbol: Zdhhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175160.3 | NP_780369.1 | 7 - 8 | 941 | 70 | |
| GenBank mRNA | AK038593.1 | - | 7 - 8 | 943 | 70 | |
| BC026570.1 | - | 3 - 4 | 522 | 4984252 | 70 | |
| BC119378.1 | - | 4 - 5 | 680 | 8734127 | 70 | |
| BC144878.1 | - | 4 - 5 | 680 | 9053426 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc1
Genomic Map