Assay Details
| Gene Symbol: Ctcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181322.3 | NP_851839.1 | 4 - 5 | 1258 | 62 | |
| GenBank mRNA | AB221631.1 | - | 2 - 3 | 949 | 62 | |
| AK132514.1 | - | 2 - 3 | 539 | 62 | ||
| AK134247.1 | - | 2 - 3 | 539 | 62 | ||
| AK140164.1 | - | 4 - 5 | 1246 | 62 | ||
| BC046398.1 | - | 3 - 4 | 982 | 3982679 | 62 | |
| BC049131.1 | - | 4 - 5 | 1217 | 6825952 | 62 | |
| BC058240.1 | - | 3 - 4 | 1011 | 6307925 | 62 | |
| U51037.1 | - | 4 - 5 | 1255 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctcf
Genomic Map