Assay Details
| Gene Symbol: Ctcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181322.3 | NP_851839.1 | 7 - 8 | 1664 | 121 | |
| GenBank mRNA | AB221631.1 | - | 5 - 6 | 1355 | 121 | |
| AK132514.1 | - | 5 - 6 | 945 | 121 | ||
| AK134247.1 | - | 5 - 6 | 945 | 121 | ||
| AK140164.1 | - | 7 - 8 | 1652 | 121 | ||
| BC046398.1 | - | 6 - 7 | 1388 | 3982679 | 121 | |
| BC049131.1 | - | 7 - 8 | 1623 | 6825952 | 121 | |
| BC058240.1 | - | 6 - 7 | 1417 | 6307925 | 121 | |
| U51037.1 | - | 7 - 8 | 1661 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctcf
Genomic Map