Assay Details
| Gene Symbol: Cacng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019431.2 | NP_062304.1 | 1 - 2 | 218 | 77 | |
| GenBank mRNA | AK144253.1 | - | 1 - 2 | 449 | 77 | |
| BC138503.1 | - | 1 - 2 | 237 | 8861523 | 77 | |
| BC138506.1 | - | 1 - 2 | 237 | 8861526 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cacng4
Genomic Map