Assay Details
| Gene Symbol: Alkbh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026303.1 | NP_080579.1 | 2 - 3 | 154 | 61 | |
| GenBank mRNA | AK020197.1 | - | 2 - 3 | 257 | 61 | |
| AK033352.1 | - | 2 - 3 | 393 | 61 | ||
| AK041810.1 | - | 2 - 3 | 245 | 61 | ||
| AK054267.1 | - | 2 - 3 | 406 | 61 | ||
| AK081459.1 | - | 2 - 3 | 406 | 61 | ||
| AK133489.1 | - | 2 - 3 | 252 | 61 | ||
| AK160783.1 | - | 2 - 3 | 246 | 61 | ||
| BC050863.1 | - | 2 - 3 | 154 | 6329366 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK081459.1 transcripts.
Set Membership: |
|

Alkbh8
Genomic Map