Assay Details
| Gene Symbol: Alkbh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026303.1 | NP_080579.1 | 6 - 7 | 727 | 125 | |
| GenBank mRNA | AK033352.1 | - | 6 - 7 | 965 | 124 | |
| AK041810.1 | - | 6 - 7 | 818 | 125 | ||
| AK054267.1 | - | 7 - 8 | 1127 | 125 | ||
| AK081459.1 | - | 6 - 7 | 978 | 125 | ||
| AK133489.1 | - | 6 - 7 | 825 | 125 | ||
| BC050863.1 | - | 6 - 7 | 727 | 6329366 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alkbh8
Genomic Map