Assay Details
| Gene Symbol: Alkbh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026303.1 | NP_080579.1 | 7 - 8 | 799 | 82 | |
| GenBank mRNA | AK033352.1 | - | 7 - 8 | 1037 | 82 | |
| AK041810.1 | - | 7 - 8 | 890 | 82 | ||
| AK054267.1 | - | 8 - 9 | 1199 | 82 | ||
| AK081459.1 | - | 7 - 8 | 1050 | 82 | ||
| AK160783.1 | - | 6 - 7 | 786 | 82 | ||
| BC050863.1 | - | 7 - 8 | 799 | 6329366 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alkbh8
Genomic Map