Assay Details
| Gene Symbol: Alkbh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026303.1 | NP_080579.1 | 9 - 10 | 1055 | 74 | |
| GenBank mRNA | AK033352.1 | - | 9 - 10 | 1293 | 74 | |
| AK041810.1 | - | 9 - 10 | 1146 | 74 | ||
| AK054267.1 | - | 10 - 11 | 1455 | 74 | ||
| AK081459.1 | - | 9 - 10 | 1306 | 74 | ||
| AK133489.1 | - | 9 - 10 | 1151 | 74 | ||
| AK160783.1 | - | 8 - 9 | 1042 | 74 | ||
| BC050863.1 | - | 9 - 10 | 1055 | 6329366 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alkbh8
Genomic Map