Assay Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028131.3 | NP_082407.1 | 11 - 12 | 1101 | 136 | |
| GenBank mRNA | BC027119.1 | - | 8 - 9 | 1825 | 136 | |
| BC058243.1 | - | 9 - 10 | 1012 | 6309162 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199702 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 395153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705318 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK010547.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK044690.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK077616.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK145598.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK145719.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK145888.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AK146274.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | AY009405.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | BC038332.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | BC050787.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853626 | NM_025680.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106188 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123830 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26199807 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06655426 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123830 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106188 | - | - |
| Whole Mouse Genome G4122A | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | BC027119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | BC058243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | NM_028131.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P304947 | AK012097.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109814 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123830 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106188 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06857259 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06857267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06857257 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06857256 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26199704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26199763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07016726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07232869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06845424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06857271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06857259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06977071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06853654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07238853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07104572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06791496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26199805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06891389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575744 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427105_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452305_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575733 | - | - |
| Mouse Genome 430 2.0 Array | 1452305_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1452305_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427105_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427105_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72155_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72155_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011311.mm.1 | NM_028131.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011311.mm.1 | NM_028131.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011311.mm.1 | TC0800001390.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452305_s_at | - | - |
| Mouse Expression Set 430 | 1427105_PM_at | - | - |
| Mouse Expression Set 430 | 1452305_PM_s_at | - | - |
| Mouse Expression Set 430 | 1427105_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427105_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452305_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452305_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427105_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427105_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452305_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427105_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452305_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 513334 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5390500 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5390500 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005390500 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223979 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223979 | - | - |
| mouse-6 v1.1 (Illumina) | scl33279.12.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5390500 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33279.12.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5390500 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223979 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2897468 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2897468 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223979 | - | - |

Cenpn
Genomic Map