Assay Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028131.3 | NP_082407.1 | 4 - 5 | 389 | 95 | |
| GenBank mRNA | AK012097.1 | - | 4 - 5 | 390 | 95 | |
| BC027119.1 | - | 2 - 3 | 204 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpn
Genomic Map