Assay Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028131.3 | NP_082407.1 | 5 - 6 | 444 | 141 | |
| GenBank mRNA | AK012097.1 | - | 5 - 6 | 445 | 141 | |
| BC027119.1 | - | 3 - 4 | 259 | 141 | ||
| BC058243.1 | - | 3 - 4 | 355 | 6309162 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058243.1 transcripts.
Set Membership: |
|

Cenpn
Genomic Map