Assay Details
| Gene Symbol: Eif2s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026030.2 | NP_080306.1 | 6 - 7 | 885 | 115 | |
| GenBank mRNA | AK012338.1 | - | 6 - 7 | 796 | 115 | |
| AK012817.1 | - | 5 - 6 | 675 | 115 | ||
| AK145401.1 | - | 6 - 7 | 790 | 115 | ||
| AK145434.1 | - | 6 - 7 | 832 | 115 | ||
| AK166862.1 | - | 6 - 7 | 789 | 115 | ||
| BC003848.1 | - | 6 - 7 | 761 | 3594058 | 115 | |
| BC100539.1 | - | 6 - 7 | 797 | 115 | ||
| BC113766.1 | - | 6 - 7 | 757 | 40058503 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif2s2
Genomic Map