Assay Details
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012392.1 | NP_001012392.1 | 10 - 11 | 1103 | 91 | |
NM_153418.2 | NP_700467.2 | 10 - 11 | 1066 | 91 | ||
GenBank mRNA | AK146605.1 | - | 10 - 11 | 1099 | 91 | |
BC060982.1 | - | 10 - 11 | 1103 | 30300112 | 91 | |
BC100436.1 | - | 10 - 11 | 1098 | 30298932 | 91 | |
U46068.3 | - | 10 - 11 | 1067 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
