Assay Details
| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021534.3 | NP_067509.1 | 1 - 2 | 150 | 86 | |
| GenBank mRNA | AJ271476.1 | - | 1 - 2 | 139 | 86 | |
| AK003127.1 | - | 1 - 2 | 165 | 86 | ||
| AK019381.1 | - | 1 - 2 | 165 | 86 | ||
| AK050647.1 | - | 1 - 2 | 151 | 86 | ||
| AK143785.1 | - | 1 - 2 | 143 | 86 | ||
| AK156743.1 | - | 150 | 86 | |||
| BC025796.1 | - | 1 - 2 | 129 | 5100832 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pxmp4
Genomic Map