Assay Details
| Gene Symbol: Tox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172913.3 | NP_766501.2 | 5 - 6 | 1200 | 90 | |
| GenBank mRNA | AK082602.1 | - | 5 - 6 | 1198 | 90 | |
| BC052044.1 | - | 5 - 6 | 926 | 5708447 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox3
Genomic Map