Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019307.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019307.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019307.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019307.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019307.mm.1 |
TC0200002396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019314.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019314.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019314.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019314.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019314.mm.1 |
TC0200002396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019308.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019308.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019308.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019308.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019308.mm.1 |
TC0200002396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019315.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019315.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019315.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019315.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019315.mm.1 |
TC0200002396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019313.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019313.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019313.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019313.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019313.mm.1 |
TC0200002396.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019309.mm.1 |
NM_001164818.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019309.mm.1 |
NM_001164818.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019309.mm.1 |
NM_026661.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019309.mm.1 |
NM_026661.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019309.mm.1 |
TC0200002396.mm.1 |
MTA TC |