Assay Details
| Gene Symbol: Sla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF287467.1 | - | 1 - 2 | 239 | 63 | |
| AF434990.1 | - | 1 - 2 | 231 | 63 | ||
| AK088672.1 | - | 1 - 2 | 315 | 63 | ||
| BC052655.1 | - | 1 - 2 | 287 | 30040401 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF287467.1, AF434990.1, AK088672.1, BC052655.1 transcripts.
Set Membership: |
|

Sla2
Genomic Map