Assay Details
| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025516.4 | NP_079792.1 | 8 - 9 | 730 | 82 | |
| GenBank mRNA | AK009350.1 | - | 8 - 9 | 731 | 82 | |
| AK013942.1 | - | 8 - 9 | 730 | 82 | ||
| AK050285.1 | - | 8 - 9 | 745 | 82 | ||
| AK183374.1 | - | 3 - 4 | 187 | 82 | ||
| AK190333.1 | - | 4 - 5 | 306 | 82 | ||
| BC043720.1 | - | 8 - 9 | 755 | 5375947 | 82 | |
More Information
Set Membership: |
|

Ergic3
Genomic Map