Assay Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027946.3 | NP_082222.1 | 2 - 3 | 547 | 62 | |
| GenBank mRNA | AK005902.1 | - | 2 - 3 | 541 | 62 | |
| AK047435.1 | - | 2 - 3 | 549 | 62 | ||
| AK080823.1 | - | 2 - 3 | 520 | 62 | ||
| AK154757.1 | - | 2 - 3 | 538 | 62 | ||
| AK165674.1 | - | 2 - 3 | 538 | 62 | ||
| BB646282.1 | - | 2 - 3 | 548 | 62 | ||
| BC048165.1 | - | 2 - 3 | 463 | 6409531 | 62 | |
| BC048722.1 | - | 2 - 3 | 466 | 6504571 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf7
Genomic Map