Assay Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027946.3 | NP_082222.1 | 4 - 5 | 773 | 96 | |
| GenBank mRNA | AK005902.1 | - | 4 - 5 | 767 | 96 | |
| AK047435.1 | - | 4 - 5 | 775 | 96 | ||
| AK154757.1 | - | 4 - 5 | 764 | 96 | ||
| AK165674.1 | - | 4 - 5 | 764 | 96 | ||
| BC048722.1 | - | 4 - 5 | 692 | 6504571 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf7
Genomic Map