Assay Details
| Gene Symbol: Dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027946.3 | NP_082222.1 | 5 - 6 | 991 | 83 | |
| GenBank mRNA | AK005902.1 | - | 5 - 6 | 985 | 83 | |
| AK047435.1 | - | 5 - 6 | 993 | 83 | ||
| AK165674.1 | - | 5 - 6 | 982 | 83 | ||
| BC048165.1 | - | 5 - 6 | 904 | 6409531 | 83 | |
| BC048722.1 | - | 5 - 6 | 910 | 6504571 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf7
Genomic Map