Assay Details
| Gene Symbol: Efcab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081046.1 | NP_001074515.1 | 2 - 3 | 371 | 94 | |
| GenBank mRNA | AK014861.1 | - | 2 - 3 | 365 | 94 | |
| AK162207.1 | - | 11 - 12 | 856 | 94 | ||
| BC050801.1 | - | 2 - 3 | 369 | 6770929 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab3
Genomic Map