Assay Details
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081046.1 | NP_001074515.1 | 3 - 4 | 448 | 109 | |
GenBank mRNA | AK014861.1 | - | 3 - 4 | 442 | 109 | |
AK162207.1 | - | 12 - 13 | 933 | 109 | ||
BC050801.1 | - | 3 - 4 | 446 | 6770929 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
