Assay Details
| Gene Symbol: Nacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025788.3 | NP_080064.3 | 2 - 3 | 1097 | 87 | |
| GenBank mRNA | AK012935.1 | - | 2 - 3 | 1080 | 87 | |
| BC048157.1 | - | 2 - 3 | 1070 | 5697941 | 87 | |
| BC052706.1 | - | 2 - 3 | 1080 | 5702150 | 87 | |
| BC054800.1 | - | 2 - 3 | 1097 | 6850337 | 87 | |
More Information
Set Membership: |
|

Nacc1
Genomic Map