Assay Details
| Gene Symbol: Il27ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053005.1 | - | 4 - 5 | 524 | 66 | |
| AF265243.1 | - | 4 - 5 | 535 | 66 | ||
| BC032878.1 | - | 4 - 5 | 556 | 1244998 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il27ra
Genomic Map