Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 13 - 14 | 2052 | 69 | |
| GenBank mRNA | AK037226.1 | - | 6 - 7 | 995 | 69 | |
| AK083571.1 | - | 13 - 14 | 2023 | 69 | ||
| AK147790.1 | - | 13 - 14 | 2057 | 69 | ||
| BC057018.1 | - | 13 - 14 | 2031 | 5705637 | 69 | |
| X76088.1 | - | 13 - 14 | 1959 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx1
Genomic Map