Assay Details
| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145970.1 | NP_666082.2 | 20 - 21 | 2148 | 78 | |
| GenBank mRNA | BC016188.1 | - | 20 - 21 | 2148 | 4482793 | 78 |
| BC027028.1 | - | 19 - 20 | 1963 | 5363547 | 78 | |
| BC030439.1 | - | 4 - 5 | 255 | 78 | ||
| DQ329239.1 | - | 13 - 14 | 1198 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cc2d1a
Genomic Map