Assay Details
| Gene Symbol: Kcns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008435.2 | NP_032461.2 | 2 - 3 | 274 | 60 | |
| GenBank mRNA | AF008573.1 | - | 2 - 3 | 251 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF008573.1, NM_008435.2 transcripts.
Set Membership: |
|

Kcns1
Genomic Map