Assay Details
| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114079.2 | NP_001107551.1 | 1 - 2 | 448 | 89 | |
| GenBank mRNA | BC158030.1 | - | 1 - 2 | 448 | 9007333 | 89 |
| EU669908.1 | - | 1 - 2 | 191 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pabpc1l
Genomic Map