Assay Details
| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114079.2 | NP_001107551.1 | 4 - 5 | 899 | 87 | |
| GenBank mRNA | BC158030.1 | - | 4 - 5 | 899 | 9007333 | 87 |
| EU669908.1 | - | 4 - 5 | 642 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pabpc1l
Genomic Map