Assay Details
| Gene Symbol: Lgr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010195.2 | NP_034325.2 | 11 - 12 | 1358 | 69 | |
| GenBank mRNA | AF110818.1 | - | 11 - 12 | 1071 | 69 | |
| AK132387.1 | - | 11 - 12 | 1358 | 69 | ||
More Information
Set Membership: |
|

Lgr5
Genomic Map