Assay Details
| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138585.3 | NP_613051.3 | 8 - 9 | 1304 | 123 | |
| GenBank mRNA | AF536541.1 | - | 8 - 9 | 1206 | 123 | |
| AK049765.1 | - | 8 - 9 | 1214 | 123 | ||
| AK144328.1 | - | 8 - 9 | 1222 | 123 | ||
| AK171907.1 | - | 1 - 2 | 280 | 123 | ||
| AK206293.1 | - | 1 - 2 | 276 | 123 | ||
| AK212479.1 | - | 1 - 2 | 278 | 123 | ||
| AK220122.1 | - | 2 - 3 | 332 | 123 | ||
| BC032191.1 | - | 8 - 9 | 1222 | 5322208 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cherp
Genomic Map