Assay Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016776.2 | NP_058056.2 | 21 - 22 | 3011 | 59 | |
| GenBank mRNA | AK083908.1 | - | 21 - 22 | 3037 | 59 | |
| AK143469.1 | - | 21 - 22 | 3036 | 59 | ||
| AK143487.1 | - | 21 - 22 | 3044 | 59 | ||
| AK149747.1 | - | 16 - 17 | 2320 | 59 | ||
| AK154016.1 | - | 19 - 20 | 3230 | 59 | ||
| AK155074.1 | - | 21 - 22 | 3022 | 59 | ||
| AK190125.1 | - | 1 - 2 | 95 | 59 | ||
| AK198759.1 | - | 1 - 2 | 95 | 59 | ||
| BC004078.1 | - | 11 - 12 | 1495 | 59 | ||
| BC015070.1 | - | 21 - 22 | 3012 | 59 | ||
| BC048858.1 | - | 21 - 22 | 3011 | 5363740 | 59 | |
| BC052889.1 | - | 21 - 22 | 3032 | 30057169 | 59 | |
| BC060167.1 | - | 20 - 21 | 3107 | 59 | ||
| U63648.1 | - | 21 - 22 | 3020 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mybbp1a
Genomic Map