Assay Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016776.2 | NP_058056.2 | 22 - 23 | 3078 | 80 | |
| GenBank mRNA | AK083908.1 | - | 22 - 23 | 3104 | 80 | |
| AK143469.1 | - | 22 - 23 | 3103 | 80 | ||
| AK143487.1 | - | 22 - 23 | 81 | |||
| AK149747.1 | - | 17 - 18 | 2387 | 80 | ||
| AK154016.1 | - | 20 - 21 | 3297 | 80 | ||
| AK155074.1 | - | 22 - 23 | 3089 | 80 | ||
| AK166533.1 | - | 11 - 12 | 1278 | 80 | ||
| AK183304.1 | - | 2 - 3 | 162 | 80 | ||
| BC004078.1 | - | 12 - 13 | 1562 | 80 | ||
| BC015070.1 | - | 22 - 23 | 3079 | 80 | ||
| BC048858.1 | - | 22 - 23 | 3078 | 5363740 | 80 | |
| BC052889.1 | - | 22 - 23 | 3099 | 30057169 | 80 | |
| BC060167.1 | - | 21 - 22 | 3174 | 80 | ||
| U63648.1 | - | 22 - 23 | 3087 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mybbp1a
Genomic Map